Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mob1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033891.1 | NP_001029063.1 | 4 - 5 | 584 | 149 | |
GenBank mRNA | BC101879.1 | - | 4 - 5 | 584 | 7929996 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |