Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025422.1 | NP_001020593.1 | 2 - 3 | 311 | 94 | |
XM_008763194.1 | XP_008761416.1 | 2 - 3 | 355 | 94 | ||
GenBank mRNA | BC098060.1 | - | 2 - 3 | 311 | 7458113 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |