Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013095.3 | NP_037227.1 | 5 - 6 | 858 | 96 | |
XM_008766216.1 | XP_008764438.1 | 3 - 4 | 797 | 96 | ||
XM_008766217.1 | XP_008764439.1 | 3 - 4 | 523 | 96 | ||
GenBank mRNA | BC064437.1 | - | 858 | 5599910 | 96 | |
DQ409172.1 | - | 2 - 3 | 114 | 96 | ||
U66479.1 | - | 5 - 6 | 667 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |