Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107167.1 | NP_001100637.1 | 5 - 6 | 594 | 98 | |
XM_006249632.2 | XP_006249694.2 | 4 - 5 | 744 | 98 | ||
XM_006249633.2 | XP_006249695.2 | 4 - 5 | 446 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |