Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201635.2 | NP_963966.1 | 6 - 7 | 846 | 54 | |
XM_006235672.2 | XP_006235734.1 | 6 - 7 | 1188 | 54 | ||
GenBank mRNA | BC100059.1 | - | 6 - 7 | 1191 | 7129117 | 54 |
X59506.1 | - | 6 - 7 | 1189 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |