Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106544.1 | NP_001100014.1 | 7 - 8 | 587 | 94 | |
GenBank mRNA | FQ213909.1 | - | 7 - 8 | 565 | 94 | |
FQ214139.1 | - | 7 - 8 | 570 | 94 | ||
FQ216824.1 | - | 7 - 8 | 570 | 94 | ||
FQ219919.1 | - | 7 - 8 | 570 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |