Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031577.1 | NP_113765.1 | 3 - 4 | 382 | 90 | |
XM_008762304.1 | XP_008760526.1 | 3 - 4 | 284 | 90 | ||
GenBank mRNA | M73486.1 | - | 3 - 4 | 382 | 90 | |
U10156.1 | - | 3 - 4 | 284 | 90 | ||
U41183.1 | - | 2 - 3 | 205 | 90 | ||
Z34092.1 | - | 3 - 4 | 284 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |