Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044244.2 | NP_001037709.1 | 3 - 4 | 398 | 72 | |
XM_006235585.2 | XP_006235647.1 | 3 - 4 | 673 | 72 | ||
GenBank mRNA | BC105768.1 | - | 3 - 4 | 399 | 7930355 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |