Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037974.1 | NP_001033063.1 | 1 - 2 | 422 | 64 | |
XM_006235526.2 | XP_006235588.1 | 1 - 2 | 1313 | 64 | ||
GenBank mRNA | DQ304564.1 | - | 1 - 2 | 422 | 64 | |
FQ223967.1 | - | 1 - 2 | 849 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |