Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025747.1 | NP_001020918.1 | 5 - 6 | 438 | 119 | |
XM_006257077.2 | XP_006257139.1 | 7 - 8 | 595 | 119 | ||
GenBank mRNA | BC097987.1 | - | 5 - 6 | 438 | 7444864 | 119 |
FQ211792.1 | - | 5 - 6 | 523 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |