Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmral1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191588.1 | NP_001178517.1 | 5 - 6 | 742 | 93 | |
XM_006245784.1 | XP_006245846.1 | 4 - 5 | 1021 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |