Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prickle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107876.1 | NP_001101346.1 | 6 - 7 | 907 | 86 | |
XM_006236912.2 | XP_006236974.1 | 6 - 7 | 1546 | 86 | ||
XM_006236913.2 | XP_006236975.1 | 6 - 7 | 5856 | 86 | ||
XM_008763124.1 | XP_008761346.1 | 6 - 7 | 1547 | 86 | ||
XM_008763125.1 | XP_008761347.1 | 6 - 7 | 885 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |