Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thnsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009658.1 | NP_001009658.1 | 5 - 6 | 860 | 83 | |
XM_006236625.2 | XP_006236687.1 | 990 | 83 | |||
XM_006236626.2 | XP_006236688.1 | 990 | 83 | |||
XM_006236627.2 | XP_006236689.1 | 796 | 83 | |||
GenBank mRNA | BC088454.1 | - | 5 - 6 | 860 | 7127050 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |