Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021696.2 | NP_067728.1 | 1 - 2 | 195 | 65 | |
XM_006249639.2 | XP_006249701.1 | 2 - 3 | 381 | 65 | ||
XM_008769495.1 | XP_008767717.1 | 2 - 3 | 278 | 65 | ||
GenBank mRNA | X64563.1 | - | 1 - 2 | 196 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Rattus norvegicus Probe spans exons |