Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034146.1 | NP_001029318.1 | 2 - 3 | 172 | 120 | |
XM_008772609.1 | XP_008770831.1 | 2 - 3 | 502 | 120 | ||
XM_008772610.1 | XP_008770832.1 | 2 - 3 | 500 | 120 | ||
GenBank mRNA | BC105892.1 | - | 2 - 3 | 172 | 7367339 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |