Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145094.2 | NP_659562.2 | 5 - 6 | 473 | 84 | |
GenBank mRNA | BC072698.1 | - | 5 - 6 | 473 | 7105732 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |