Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-Da | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008847.2 | NP_001008847.1 | 4 - 5 | 838 | 111 | |
GenBank mRNA | BC098677.1 | - | 4 - 5 | 804 | 7373701 | 111 |
FQ218574.1 | - | 4 - 5 | 829 | 111 | ||
FQ220763.1 | - | 4 - 5 | 826 | 111 | ||
FQ220966.1 | - | 4 - 5 | 831 | 111 | ||
FQ227948.1 | - | 4 - 5 | 831 | 111 | ||
FQ230207.1 | - | 4 - 5 | 829 | 111 | ||
FQ232717.1 | - | 4 - 5 | 826 | 111 | ||
FQ232757.1 | - | 3 - 4 | 593 | 111 | ||
FQ233042.1 | - | 4 - 5 | 829 | 111 | ||
FQ233943.1 | - | 4 - 5 | 829 | 111 | ||
FQ234684.1 | - | 4 - 5 | 832 | 111 | ||
FQ235025.1 | - | 4 - 5 | 821 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |