Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256146.2 | XP_006256208.2 | 1 - 2 | 415 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Inventoried Probe spans exons |