Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191719.1 | NP_001178648.1 | 24 - 25 | 7361 | 132 | |
XM_006256359.2 | XP_006256421.1 | 24 - 25 | 7415 | 132 | ||
XM_006256360.2 | XP_006256422.1 | 23 - 24 | 7211 | 132 | ||
XM_006256361.2 | XP_006256423.1 | 24 - 25 | 7438 | 132 | ||
XM_006256362.2 | XP_006256424.1 | 24 - 25 | 7330 | 132 | ||
XM_006256363.1 | XP_006256425.1 | 20 - 21 | 6772 | 132 | ||
XM_006256364.1 | XP_006256426.1 | 24 - 25 | 7324 | 132 | ||
XM_006256365.1 | XP_006256427.1 | 19 - 20 | 6630 | 132 | ||
GenBank mRNA | AY327507.1 | - | 4 - 5 | 549 | 132 | |
FQ235173.1 | - | 2 - 3 | 289 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |