Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-DMb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198740.2 | NP_942035.2 | 1 - 2 | 277 | 160 | |
GenBank mRNA | BC091108.1 | - | 1 - 2 | 213 | 7380292 | 160 |
FQ226840.1 | - | 1 - 2 | 266 | 160 | ||
FQ227160.1 | - | 1 - 2 | 214 | 160 | ||
FQ233093.1 | - | 1 - 2 | 214 | 160 | ||
FQ233373.1 | - | 1 - 2 | 229 | 160 | ||
KC222899.1 | - | 1 - 2 | 58 | 160 | ||
KC222901.1 | - | 1 - 2 | 58 | 160 |
Gene Symbol: LOC100909630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751925.3 | XP_003751973.1 | 1 - 2 | 314 | 160 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |