Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107619.1 | NP_001101089.1 | 2 - 3 | 263 | 143 | |
XM_006256210.2 | XP_006256272.1 | 3 - 4 | 353 | 143 | ||
XM_006256211.2 | XP_006256273.1 | 3 - 4 | 356 | 143 | ||
XM_008772817.1 | XP_008771039.1 | 2 - 3 | 745 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Rattus norvegicus Probe spans exons |