Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhipl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_343107.7 | XP_343108.4 | 8 - 9 | 2333 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |