Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053341.1 | NP_445793.1 | 5 - 6 | 907 | 74 | |
XM_006255311.2 | XP_006255373.1 | 5 - 6 | 3172 | 74 | ||
XM_006255312.2 | XP_006255374.1 | 6 - 7 | 2166 | 74 | ||
XM_008772420.1 | XP_008770642.1 | 6 - 7 | 2096 | 74 | ||
GenBank mRNA | AF032120.1 | - | 5 - 6 | 907 | 74 | |
BC061964.1 | - | 4 - 5 | 1715 | 74 | ||
BC070505.1 | - | 6 - 7 | 950 | 7097697 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |