Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133581.1 | NP_598265.1 | 2 - 3 | 362 | 106 | |
XM_006255705.2 | XP_006255767.1 | 2 - 3 | 615 | 106 | ||
GenBank mRNA | AF037272.1 | - | 2 - 3 | 362 | 106 | |
BC063152.1 | - | 2 - 3 | 365 | 6887007 | 106 | |
FQ213083.1 | - | 2 - 3 | 347 | 106 | ||
FQ216345.1 | - | 2 - 3 | 356 | 106 | ||
FQ216415.1 | - | 2 - 3 | 357 | 106 | ||
FQ219846.1 | - | 2 - 3 | 357 | 106 | ||
FQ220334.1 | - | 2 - 3 | 594 | 106 | ||
FQ229404.1 | - | 2 - 3 | 347 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |