Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Banp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106191.1 | NP_001099661.1 | 3 - 4 | 298 | 105 | |
XM_006255751.2 | XP_006255813.1 | 3 - 4 | 344 | 105 | ||
XM_006255752.2 | XP_006255814.1 | 3 - 4 | 343 | 105 | ||
XM_006255753.2 | XP_006255815.1 | 3 - 4 | 343 | 105 | ||
XM_006255754.2 | XP_006255816.1 | 3 - 4 | 343 | 105 | ||
XM_006255755.2 | XP_006255817.1 | 3 - 4 | 343 | 105 | ||
XM_006255756.2 | XP_006255818.1 | 3 - 4 | 343 | 105 | ||
XM_006255757.2 | XP_006255819.1 | 3 - 4 | 342 | 105 | ||
XM_008772601.1 | XP_008770823.1 | 3 - 4 | 565 | 105 | ||
XM_008772602.1 | XP_008770824.1 | 3 - 4 | 344 | 105 | ||
GenBank mRNA | BC160844.1 | - | 3 - 4 | 325 | 8361869 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |