Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255657.2 | XP_006255719.1 | 1 - 2 | 868 | 154 | |
XM_006255660.2 | XP_006255722.1 | 1 - 2 | 867 | 121 | ||
XM_008772585.1 | XP_008770807.1 | 1 - 2 | 874 | 294 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |