Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022962.1 | NP_075251.1 | 13 - 14 | 2659 | 80 | |
GenBank mRNA | AF081144.1 | - | 12 - 13 | 2644 | 80 | |
AF081145.1 | - | 12 - 13 | 2644 | 80 | ||
AF081146.1 | - | 13 - 14 | 2659 | 80 | ||
AF081147.1 | - | 13 - 14 | 2659 | 80 | ||
AF111099.1 | - | 14 - 15 | 2985 | 80 | ||
U72487.1 | - | 12 - 13 | 2461 | 80 | ||
U78105.1 | - | 12 - 13 | 2786 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |