Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109611.1 | NP_001103081.1 | 2 - 3 | 314 | 101 | |
XM_006255634.2 | XP_006255696.1 | 2 - 3 | 1154 | 101 | ||
XM_006255635.2 | XP_006255697.1 | 2 - 3 | 1280 | 101 | ||
XM_006255636.2 | XP_006255698.1 | 2 - 3 | 1108 | 101 | ||
XM_008772535.1 | XP_008770757.1 | 2 - 3 | 1154 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |