Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107434.1 | NP_001100904.1 | 8 - 9 | 1475 | 106 | |
XM_006255685.2 | XP_006255747.1 | 8 - 9 | 2257 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |