Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctcf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031824.1 | NP_114012.1 | 9 - 10 | 1867 | 78 | |
XM_006255536.1 | XP_006255598.1 | 8 - 9 | 1817 | 78 | ||
XM_008772536.1 | XP_008770758.1 | 7 - 8 | 1834 | 78 | ||
XM_008772537.1 | XP_008770759.1 | 7 - 8 | 1834 | 78 | ||
XM_008772538.1 | XP_008770760.1 | 7 - 8 | 1834 | 78 | ||
XM_008772539.1 | XP_008770761.1 | 7 - 8 | 1834 | 78 | ||
GenBank mRNA | AF133731.2 | - | 9 - 10 | 1903 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |