Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nip7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138847.1 | NP_620202.1 | 2 - 3 | 233 | 56 | |
GenBank mRNA | AF158186.1 | - | 2 - 3 | 233 | 56 | |
BC059114.1 | - | 2 - 3 | 204 | 6921568 | 56 | |
BC086589.1 | - | 2 - 3 | 156 | 7309991 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |