Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012164.1 | NP_001012164.1 | 12 - 13 | 1458 | 69 | |
XM_006255278.2 | XP_006255340.1 | 12 - 13 | 2125 | 69 | ||
XM_006255279.2 | XP_006255341.1 | 11 - 12 | 1990 | 69 | ||
XM_006255280.1 | XP_006255342.1 | 13 - 14 | 1707 | 69 | ||
XM_006255281.2 | XP_006255343.1 | 10 - 11 | 1843 | 69 | ||
GenBank mRNA | BC083857.1 | - | 12 - 13 | 1458 | 7125718 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Rattus norvegicus Probe spans exons |