Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017511.1 | NP_001017511.1 | 3 - 4 | 1236 | 78 | |
XM_006255718.2 | XP_006255780.1 | 3 - 4 | 1348 | 78 | ||
XM_008772641.1 | XP_008770863.1 | 3 - 4 | 2598 | 78 | ||
GenBank mRNA | BC081914.1 | - | 3 - 4 | 1236 | 7112173 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |