Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238440.2 | XP_006238502.1 | 5 - 6 | 1159 | 119 | |
GenBank mRNA | FQ210243.1 | - | 5 - 6 | 985 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus Probe spans exons |