Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238423.2 | XP_006238485.1 | 27 - 28 | 3923 | 94 | |
XM_006238424.2 | XP_006238486.1 | 27 - 28 | 3929 | 94 | ||
XM_006238425.2 | XP_006238487.1 | 2 - 3 | 965 | 94 | ||
XM_008763916.1 | XP_008762138.1 | 3 - 4 | 1019 | 94 | ||
GenBank mRNA | FQ212190.1 | - | 3 - 4 | 640 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |