Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007740.1 | NP_001007741.1 | 1 - 2 | 87 | 109 | |
GenBank mRNA | BC079150.1 | - | 1 - 2 | 87 | 7128580 | 109 |
FQ211877.1 | - | 1 - 2 | 100 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |