Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256434.1 | XP_006256496.1 | 1 - 2 | 115 | 126 | |
GenBank mRNA | FQ209472.1 | - | 1 - 2 | 118 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |