Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108533.2 | NP_001102003.1 | 5 - 6 | 479 | 134 | |
XM_006256419.2 | XP_006256481.1 | 5 - 6 | 1120 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |