Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022176.2 | NP_071512.2 | 3 - 4 | 632 | 128 | |
GenBank mRNA | BC158563.1 | - | 3 - 4 | 632 | 7443132 | 128 |
FQ219971.1 | - | 3 - 4 | 648 | 128 | ||
FQ221549.1 | - | 3 - 4 | 649 | 128 | ||
FQ230053.1 | - | 3 - 4 | 129 | |||
M31155.1 | - | 3 - 4 | 591 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |