Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137626.1 | NP_001131098.1 | 5 - 6 | 1134 | 168 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Inventoried Probe spans exons |