Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam50b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107349.1 | NP_001100819.1 | 1 - 2 | 153 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |