Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013107.1 | NP_037239.1 | 5 - 6 | 1259 | 140 | |
GenBank mRNA | AY184240.1 | - | 5 - 6 | 1259 | 140 | |
X58830.1 | - | 362 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |