Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253671.2 | XP_006253733.1 | 1 - 2 | 61 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |