Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144745.2 | NP_653346.2 | 7 - 8 | 1083 | 72 | |
XM_006233670.2 | XP_006233732.1 | 7 - 8 | 1187 | 72 | ||
GenBank mRNA | AF107723.1 | - | 6 - 7 | 1207 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |