Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdccag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013135.2 | NP_001013153.2 | 3 - 4 | 390 | 65 | |
XM_006233682.2 | XP_006233744.1 | 2 - 3 | 219 | 65 | ||
XM_006233683.2 | XP_006233745.1 | 3 - 4 | 279 | 65 | ||
XM_006233686.2 | XP_006233748.1 | 3 - 4 | 369 | 65 |
Gene Symbol: RGD1562289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_576917.5 | XP_576917.1 | 1 | 224 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |