Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107834.1 | NP_001101304.1 | 1 - 2 | 252 | 92 | |
XM_006233975.2 | XP_006234037.1 | 1 - 2 | 314 | 92 |
Gene Symbol: LOC102551296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234031.2 | XP_006234093.1 | 1 - 2 | 264 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons |