Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5kl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749451.3 | XP_003749499.2 | 2 - 3 | 1859 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |