Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106555.1 | NP_001100025.1 | 5 - 6 | 609 | 70 | |
XM_006233581.2 | XP_006233643.1 | 5 - 6 | 994 | 70 | ||
GenBank mRNA | FQ212796.1 | - | 4 - 5 | 748 | 70 | |
FQ218819.1 | - | 5 - 6 | 569 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |