Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023094.3 | NP_075582.3 | 1 - 2 | 87 | 73 | |
GenBank mRNA | AF264018.2 | - | 1 - 2 | 87 | 73 | |
AF469945.1 | - | 1 - 2 | 87 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |