Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013164.3 | NP_001013182.3 | 12 - 13 | 1316 | 76 | |
XM_006233810.2 | XP_006233872.1 | 12 - 13 | 1693 | 76 | ||
XM_006233811.2 | XP_006233873.1 | 12 - 13 | 1786 | 76 | ||
XM_006233812.2 | XP_006233874.1 | 12 - 13 | 1244 | 76 | ||
XM_006233813.2 | XP_006233875.2 | 12 - 13 | 1256 | 76 | ||
XM_008761658.1 | XP_008759880.1 | 13 - 14 | 1999 | 76 | ||
GenBank mRNA | S61960.1 | - | 12 - 13 | 1457 | 76 | |
Z49696.1 | - | 13 - 14 | 1465 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |